Assay Details
| Gene Symbol: myoz1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002447.1 | NP_001002447.1 | 3 - 4 | 316 | 67 | |
| XM_005156283.4 | XP_005156340.1 | 3 - 4 | 434 | 67 | ||
| XM_005156285.3 | XP_005156342.1 | 3 - 4 | 442 | 67 | ||
| XM_017358446.2 | XP_017213935.1 | 3 - 4 | 358 | 67 | ||
| GenBank mRNA | BC076024.1 | - | 3 - 4 | 316 | 67 | |
| BC152175.1 | - | 3 - 4 | 412 | 67 | ||
More Information
Set Membership: |
|

myoz1b
Genomic Map