Assay Details
| Gene Symbol: afap1l1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030213.1 | NP_001025384.1 | 17 - 18 | 2189 | 151 | |
| XM_005173256.4 | XP_005173313.1 | 17 - 18 | 2517 | 151 | ||
| XM_009291126.3 | XP_009289401.1 | 17 - 18 | 2515 | 151 | ||
| GenBank mRNA | BC097145.1 | - | 17 - 18 | 2189 | 151 | |
More Information
Set Membership: |
|

afap1l1a
Genomic Map