Assay Details
| Gene Symbol: dkc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033107.2 | NP_001028279.2 | 14 - 15 | 1523 | 101 | |
| GenBank mRNA | BC100010.1 | - | 14 - 15 | 1487 | 104 | |
More Information
Set Membership: |
|

dkc1
Genomic Map