Assay Details
| Gene Symbol: mrps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033596.1 | NP_001028768.1 | 4 - 5 | 491 | 64 | |
| GenBank mRNA | BC100013.1 | - | 4 - 5 | 491 | 64 | |
More Information
Set Membership: |
|

mrps7
Genomic Map