Assay Details
| Gene Symbol: zgc:123238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037112.1 | NP_001032189.1 | 1 - 2 | 228 | 97 | |
| GenBank mRNA | BC107520.1 | - | 1 - 2 | 228 | 97 | |
| BC171453.1 | - | 1 - 2 | 205 | 97 | ||
| BC171479.1 | - | 1 - 2 | 205 | 97 | ||
| Gene Symbol: LOC110438453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021470918.1 | XP_021326593.1 | 1 - 2 | 237 | 97 | |
More Information
Set Membership: |
|

zgc:123238
Genomic Map