Assay Details
| Gene Symbol: itgb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037235.1 | NP_001032312.1 | 4 - 5 | 665 | 55 | |
| GenBank mRNA | BC163266.1 | - | 4 - 5 | 635 | 55 | |
| DQ178647.1 | - | 4 - 5 | 665 | 55 | ||
More Information
Set Membership: |
|

itgb3a
Genomic Map