Assay Details
| Gene Symbol: actn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037573.1 | NP_001032662.1 | 11 - 12 | 1416 | 88 | |
| GenBank mRNA | BC110103.1 | - | 11 - 12 | 1416 | 88 | |
| FJ979800.1 | - | 11 - 12 | 1281 | 88 | ||
More Information
Set Membership: |
|

actn2b
Genomic Map