Assay Details
| Gene Symbol: actn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037573.1 | NP_001032662.1 | 14 - 15 | 1816 | 73 | |
| GenBank mRNA | BC110103.1 | - | 14 - 15 | 1816 | 73 | |
More Information
Set Membership: |
|

actn2b
Genomic Map