Assay Details
| Gene Symbol: actn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037573.1 | NP_001032662.1 | 6 - 7 | 786 | 110 | |
| GenBank mRNA | BC110103.1 | - | 6 - 7 | 786 | 110 | |
| FJ979800.1 | - | 6 - 7 | 110 | |||
More Information
Set Membership: |
|

actn2b
Genomic Map