Assay Details
| Gene Symbol: rad23ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037687.1 | NP_001032776.1 | 7 - 8 | 896 | 93 | |
| GenBank mRNA | BC108000.1 | - | 7 - 8 | 896 | 93 | |
| BC152590.1 | - | 7 - 8 | 855 | 93 | ||
More Information
Set Membership: |
|

rad23ab
Genomic Map