Assay Details
| Gene Symbol: nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037704.2 | NP_001032793.2 | 3 - 4 | 607 | 121 | |
| GenBank mRNA | AB178827.1 | - | 3 - 4 | 298 | 121 | |
| BC124089.1 | - | 3 - 4 | 359 | 121 | ||
More Information
Set Membership: |
|

nenf
Genomic Map