Assay Details
| Gene Symbol: slc8a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039144.1 | NP_001034233.1 | 2 - 3 | 1941 | 106 | |
| GenBank mRNA | AY934776.1 | - | 2 - 3 | 1941 | 106 | |
More Information
Set Membership: |
|

slc8a1b
Genomic Map