Assay Details
| Gene Symbol: aars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040035.2 | NP_001035124.2 | 9 - 10 | 1339 | 76 | |
| GenBank mRNA | BC097014.1 | - | 9 - 10 | 76 | ||
More Information
Set Membership: |
|

aars1
Genomic Map