Assay Details
| Gene Symbol: baiap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040245.1 | NP_001035335.1 | 8 - 9 | 978 | 89 | |
| XM_005163755.4 | XP_005163812.1 | 8 - 9 | 1040 | 89 | ||
| XM_009299775.3 | XP_009298050.1 | 8 - 9 | 1041 | 89 | ||
| XM_009299776.2 | XP_009298051.1 | 8 - 9 | 1040 | 89 | ||
| XM_021471329.1 | XP_021327004.1 | 8 - 9 | 1040 | 89 | ||
| XM_021471351.1 | XP_021327026.1 | 8 - 9 | 1040 | 89 | ||
| GenBank mRNA | BC115281.1 | - | 8 - 9 | 978 | 89 | |
More Information
Set Membership: |
|

baiap2a
Genomic Map