Assay Details
| Gene Symbol: baiap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040245.1 | NP_001035335.1 | 11 - 12 | 1466 | 62 | |
| XM_005163755.4 | XP_005163812.1 | 11 - 12 | 1528 | 62 | ||
| XM_021471329.1 | XP_021327004.1 | 11 - 12 | 1528 | 62 | ||
| GenBank mRNA | BC115281.1 | - | 11 - 12 | 1466 | 62 | |
More Information
Set Membership: |
|

baiap2a
Genomic Map