Assay Details
| Gene Symbol: cirbpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040321.2 | NP_001035411.2 | 2 - 3 | 237 | 176 | |
| NM_001309458.1 | NP_001296387.1 | 2 - 3 | 237 | 176 | ||
| NR_132109.1 | 2 - 3 | 237 | 176 | |||
| NR_132111.1 | 2 - 3 | 237 | 176 | |||
| NR_132112.1 | 2 - 3 | 237 | 176 | |||
| NR_132113.1 | 2 - 3 | 237 | 176 | |||
| XM_021469377.1 | XP_021325052.1 | 2 - 3 | 241 | 176 | ||
| GenBank mRNA | BC048027.1 | - | 2 - 3 | 176 | ||
| BC115177.1 | - | 173 | 137 | |||
| BC164022.1 | - | 1 - 2 | 176 | |||
| EH436305.1 | - | 2 - 3 | 199 | 176 | ||
| EH447371.1 | - | 2 - 3 | 202 | 176 | ||
| EH452212.1 | - | 2 - 3 | 178 | 176 | ||
| EH485709.1 | - | 2 - 3 | 185 | 176 | ||
| EH507728.1 | - | 2 - 3 | 205 | 176 | ||
More Information
Set Membership: |
|

cirbpb
Genomic Map