Assay Details
| Gene Symbol: cirbpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040321.2 | NP_001035411.2 | 5 - 6 | 678 | 58 | |
| NM_001309458.1 | NP_001296387.1 | 5 - 6 | 678 | 58 | ||
| NR_132109.1 | 5 - 6 | 678 | 58 | |||
| XM_021469377.1 | XP_021325052.1 | 5 - 6 | 682 | 58 | ||
| GenBank mRNA | BC115177.1 | - | 4 - 5 | 575 | 58 | |
| EH436305.1 | - | 5 - 6 | 641 | 58 | ||
| EH452212.1 | - | 5 - 6 | 619 | 58 | ||
More Information
Set Membership: |
|

cirbpb
Genomic Map