Assay Details
| Gene Symbol: stmn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040375.1 | NP_001035465.1 | 2 - 3 | 258 | 96 | |
| XM_021468136.1 | XP_021323811.1 | 3 - 4 | 246 | 96 | ||
| GenBank mRNA | BC115338.1 | - | 2 - 3 | 258 | 96 | |
More Information
Set Membership: |
|

stmn1a
Genomic Map