Assay Details
| Gene Symbol: top1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044324.2 | NP_001037789.1 | 18 - 19 | 2054 | 91 | |
| GenBank mRNA | BC117588.1 | - | 18 - 19 | 2054 | 91 | |
More Information
Set Membership: |
|

top1l
Genomic Map