Assay Details
| Gene Symbol: wbp1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044333.1 | NP_001037798.1 | 2 - 3 | 275 | 81 | |
| XM_021480585.1 | XP_021336260.1 | 2 - 3 | 498 | 81 | ||
| GenBank mRNA | BC117666.1 | - | 2 - 3 | 275 | 81 | |
More Information
Set Membership: |
|

wbp1la
Genomic Map