Assay Details
| Gene Symbol: rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044338.2 | NP_001037803.2 | 6 - 7 | 699 | 72 | |
| XM_009299961.3 | XP_009298236.1 | 6 - 7 | 710 | 72 | ||
| GenBank mRNA | BC133972.1 | - | 6 - 7 | 699 | 72 | |
More Information
Set Membership: |
|

rad51ap1
Genomic Map