Assay Details
| Gene Symbol: taf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044811.2 | NP_001038276.1 | 2 - 3 | 499 | 120 | |
| GenBank mRNA | BC133967.1 | - | 2 - 3 | 499 | 120 | |
More Information
Set Membership: |
|

taf10
Genomic Map