Assay Details
| Gene Symbol: taf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044811.2 | NP_001038276.1 | 3 - 4 | 554 | 130 | |
| GenBank mRNA | BC133967.1 | - | 3 - 4 | 554 | 130 | |
More Information
Set Membership: |
|

taf10
Genomic Map