Assay Details
| Gene Symbol: arl3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044908.2 | NP_001038373.1 | 2 - 3 | 184 | 72 | |
| GenBank mRNA | BC124704.1 | - | 2 - 3 | 184 | 72 | |
| BC164965.1 | - | 1 - 2 | 178 | 72 | ||
More Information
Set Membership: |
|

arl3b
Genomic Map