Assay Details
| Gene Symbol: ccnd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045068.1 | NP_001038533.1 | 4 - 5 | 715 | 65 | |
| XM_005164521.4 | XP_005164578.1 | 5 - 6 | 989 | 65 | ||
| GenBank mRNA | FJ648074.1 | - | 4 - 5 | 778 | 65 | |
More Information
Set Membership: |
|

ccnd2b
Genomic Map