Assay Details
| Gene Symbol: nab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045086.1 | NP_001038551.1 | 2 - 3 | 920 | 71 | |
| XM_005161686.4 | XP_005161743.1 | 3 - 4 | 1187 | 71 | ||
| XM_009296205.3 | XP_009294480.1 | 3 - 4 | 1190 | 71 | ||
More Information
Set Membership: |
|

nab1a
Genomic Map