Assay Details
| Gene Symbol: nab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045086.1 | NP_001038551.1 | 4 - 5 | 1086 | 131 | |
| XM_005161686.4 | XP_005161743.1 | 5 - 6 | 1353 | 131 | ||
More Information
Set Membership: |
|

nab1a
Genomic Map