Assay Details
| Gene Symbol: nab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045086.1 | NP_001038551.1 | 5 - 6 | 1251 | 61 | |
| XM_005161686.4 | XP_005161743.1 | 6 - 7 | 1518 | 61 | ||
| XM_009296205.3 | XP_009294480.1 | 7 - 8 | 1539 | 61 | ||
More Information
Set Membership: |
|

nab1a
Genomic Map