Assay Details
| Gene Symbol: nab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045086.1 | NP_001038551.1 | 6 - 7 | 1396 | 103 | |
| XM_005161686.4 | XP_005161743.1 | 7 - 8 | 1663 | 103 | ||
| XM_009296205.3 | XP_009294480.1 | 8 - 9 | 1684 | 103 | ||
More Information
Set Membership: |
|

nab1a
Genomic Map