Assay Details
| Gene Symbol: tp53bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045153.1 | NP_001038618.1 | 7 - 8 | 960 | 75 | |
| XM_005161809.4 | XP_005161866.1 | 8 - 9 | 1456 | 75 | ||
More Information
Set Membership: |
|

tp53bp2b
Genomic Map