Assay Details
| Gene Symbol: tp53bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045153.1 | NP_001038618.1 | 8 - 9 | 1147 | 80 | |
| XM_005161809.4 | XP_005161866.1 | 9 - 10 | 1643 | 80 | ||
| XM_009296445.3 | XP_009294720.1 | 9 - 10 | 1640 | 80 | ||
More Information
Set Membership: |
|

tp53bp2b
Genomic Map