Assay Details
| Gene Symbol: inppl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039135.2 | NP_001034224.2 | 3 - 4 | 397 | 61 | |
| GenBank mRNA | BC163188.1 | - | 3 - 4 | 417 | 61 | |
| DQ272662.1 | - | 3 - 4 | 370 | 61 | ||
More Information
Set Membership: |
|

inppl1b
Genomic Map