Assay Details
| Gene Symbol: inppl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039135.2 | NP_001034224.2 | 4 - 5 | 98 | ||
| GenBank mRNA | BC163188.1 | - | 4 - 5 | 98 | ||
| DQ272662.1 | - | 4 - 5 | 513 | 98 | ||
More Information
Set Membership: |
|

inppl1b
Genomic Map