Assay Details
| Gene Symbol: foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045263.2 | NP_001038728.2 | 2 - 3 | 735 | 104 | |
| XM_009292896.3 | XP_009291171.1 | 2 - 3 | 743 | 104 | ||
| XM_021467224.1 | XP_021322899.1 | 2 - 3 | 1135 | 104 | ||
| XM_021467225.1 | XP_021322900.1 | 2 - 3 | 1843 | 104 | ||
| GenBank mRNA | BC116592.1 | - | 2 - 3 | 735 | 104 | |
More Information
Set Membership: |
|

foxn3
Genomic Map