Assay Details
| Gene Symbol: rpl22l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045335.2 | NP_001038800.1 | 2 - 3 | 192 | 79 | |
| GenBank mRNA | BC118681.1 | - | 2 - 3 | 139 | 79 | |
More Information
Set Membership: |
|

rpl22l1
Genomic Map