Assay Details
| Gene Symbol: rpl22l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045335.2 | NP_001038800.1 | 3 - 4 | 310 | 93 | |
| NR_027625.1 | 3 - 4 | 378 | 93 | |||
| GenBank mRNA | BC118681.1 | - | 3 - 4 | 257 | 93 | |
| BC122324.1 | - | 3 - 4 | 323 | 93 | ||
| EH565022.1 | - | 1 - 2 | 272 | 93 | ||
More Information
Set Membership: |
|

rpl22l1
Genomic Map