Assay Details
Gene Symbol: frmd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045356.2 | NP_001038821.2 | 7 - 8 | 677 | 109 | |
XM_005173898.4 | XP_005173955.2 | 7 - 8 | 773 | 109 | ||
XM_021468906.1 | XP_021324581.1 | 4 - 5 | 556 | 109 | ||
XM_021468907.1 | XP_021324582.1 | 1 - 2 | 216 | 109 | ||
XM_021468908.1 | XP_021324583.1 | 7 - 8 | 773 | 109 | ||
GenBank mRNA | BC122375.1 | - | 7 - 8 | 610 | 109 | |
CK353164.1 | - | 7 - 8 | 627 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021468906.1, XM_021468907.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
