Assay Details
| Gene Symbol: chrna4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048063.1 | NP_001041528.1 | 3 - 4 | 444 | 90 | |
| GenBank mRNA | BC171545.1 | - | 3 - 4 | 433 | 90 | |
| DQ822508.1 | - | 3 - 4 | 444 | 90 | ||
More Information
Set Membership: |
|

chrna4b
Genomic Map