Assay Details
| Gene Symbol: hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075111.2 | NP_001068579.2 | 1 - 2 | 241 | 62 | |
| XM_021470189.1 | XP_021325864.1 | 1 - 2 | 664 | 62 | ||
| GenBank mRNA | BC124293.1 | - | 1 - 2 | 240 | 62 | |
| BC165580.1 | - | 1 - 2 | 121 | 62 | ||
| EH501708.1 | - | 1 - 2 | 274 | 62 | ||
More Information
Set Membership: |
|

hnf4g
Genomic Map