Assay Details
| Gene Symbol: prps1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076568.2 | NP_001070036.1 | 3 - 4 | 602 | 140 | |
| GenBank mRNA | BC124318.1 | - | 3 - 4 | 578 | 140 | |
| BC153527.1 | - | 3 - 4 | 524 | 140 | ||
| BC164343.1 | - | 3 - 4 | 437 | 140 | ||
More Information
Set Membership: |
|

prps1b
Genomic Map