Assay Details
| Gene Symbol: prps1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076568.2 | NP_001070036.1 | 5 - 6 | 908 | 92 | |
| GenBank mRNA | BC124318.1 | - | 5 - 6 | 884 | 92 | |
| BC153527.1 | - | 5 - 6 | 830 | 92 | ||
| BC164343.1 | - | 5 - 6 | 743 | 92 | ||
More Information
Set Membership: |
|

prps1b
Genomic Map