Assay Details
| Gene Symbol: rangap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076616.1 | NP_001070084.1 | 3 - 4 | 293 | 97 | |
| GenBank mRNA | BC124540.1 | - | 3 - 4 | 293 | 97 | |
More Information
Set Membership: |
|

rangap1a
Genomic Map