Assay Details
| Gene Symbol: ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076625.1 | NP_001070093.1 | 2 - 3 | 693 | 63 | |
| XM_005156693.4 | XP_005156750.1 | 2 - 3 | 764 | 63 | ||
| GenBank mRNA | BC124319.1 | - | 2 - 3 | 693 | 63 | |
| BC164082.1 | - | 2 - 3 | 279 | 63 | ||
| HF565571.1 | - | 2 - 3 | 245 | 63 | ||
More Information
Set Membership: |
|

ttl
Genomic Map