Assay Details
| Gene Symbol: cenatac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327927.1 | NP_001314856.1 | 4 - 5 | 534 | 99 | |
| XM_005172002.4 | XP_005172059.1 | 4 - 5 | 545 | 99 | ||
| XM_005172003.4 | XP_005172060.1 | 4 - 5 | 546 | 99 | ||
| XM_009301864.3 | XP_009300139.1 | 4 - 5 | 543 | 99 | ||
| GenBank mRNA | BC124232.1 | - | 4 - 5 | 525 | 99 | |
More Information
Set Membership: |
|

cenatac
Genomic Map