Assay Details
| Gene Symbol: cenatac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327927.1 | NP_001314856.1 | 8 - 9 | 797 | 83 | |
| XM_009301864.3 | XP_009300139.1 | 8 - 9 | 803 | 83 | ||
| GenBank mRNA | BC124232.1 | - | 7 - 8 | 961 | 83 | |
More Information
Set Membership: |
|

cenatac
Genomic Map