Assay Details
| Gene Symbol: rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076753.1 | NP_001070221.1 | 2 - 3 | 402 | 90 | |
| XM_017354591.2 | XP_017210080.1 | 2 - 3 | 486 | 90 | ||
| GenBank mRNA | BC124362.1 | - | 2 - 3 | 402 | 90 | |
More Information
Set Membership: |
|

rpusd1
Genomic Map