Assay Details
| Gene Symbol: cfap100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076760.1 | NP_001070228.1 | 13 - 14 | 1671 | 62 | |
| GenBank mRNA | BC124258.1 | - | 13 - 14 | 1671 | 62 | |
More Information
Set Membership: |
|

cfap100
Genomic Map