Assay Details
| Gene Symbol: abi3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076774.2 | NP_001070242.2 | 2 - 3 | 439 | 62 | |
| XM_005164048.4 | XP_005164105.1 | 2 - 3 | 577 | 62 | ||
| XM_021471554.1 | XP_021327229.1 | 2 - 3 | 585 | 62 | ||
| GenBank mRNA | BC124443.1 | - | 2 - 3 | 432 | 62 | |
| BC165589.1 | - | 2 - 3 | 331 | 62 | ||
More Information
Set Membership: |
|

abi3a
Genomic Map