Assay Details
| Gene Symbol: oxct1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077150.1 | NP_001070618.1 | 16 - 17 | 1593 | 77 | |
| GenBank mRNA | BC124298.1 | - | 16 - 17 | 1593 | 77 | |
| BC154254.1 | - | 16 - 17 | 1575 | 77 | ||
| BC155216.1 | - | 16 - 17 | 1615 | 77 | ||
| Gene Symbol: LOC797311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021469231.1 | XP_021324906.1 | 14 - 15 | 1261 | 77 | |
More Information
Set Membership: |
|

oxct1b
Genomic Map