Assay Details
| Gene Symbol: oxct1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077150.1 | NP_001070618.1 | 8 - 9 | 910 | 68 | |
| GenBank mRNA | BC124298.1 | - | 8 - 9 | 910 | 68 | |
| BC154254.1 | - | 8 - 9 | 892 | 68 | ||
| BC155216.1 | - | 8 - 9 | 932 | 68 | ||
| BC164688.1 | - | 8 - 9 | 894 | 68 | ||
| Gene Symbol: LOC797311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021469231.1 | XP_021324906.1 | 6 - 7 | 584 | 68 | |
More Information
Set Membership: |
|

oxct1b
Genomic Map